Learn More
Labconco SteamScrubber Culture Tube Inserts
Culture Tube Inserts for use with Labconco FlaskScrubber and SteamScrubber Glassware Washers.
$117.00 - $412.00
Catalog Number | Mfr. No. | Depth (English) | Height (English) | Weight (English) Shipping | Width (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Depth (English) | Height (English) | Weight (English) Shipping | Width (English) | Price | Quantity & Availability | ||
15-352-815
|
Labconco
4402001 |
7.8 in. | 6.4 in. | 2.5 lb. | 18 in. |
Each for $412.00
|
|
15-352-816
|
Labconco
4402101 |
8.3 in. | 9.9 in. | 2.5 lb. | 18 in. |
Each for $315.00
|
|
15-352-817
|
Labconco
4401401 |
8.4 in. | 0.3 in. | 1.5 lb. | 18.5 in. |
Each for $117.00
|
|
Description
Culture Tube Inserts for use with Labconco FlaskScrubber and SteamScrubber Glassware Washers.
Type 304 stainless steel basket-style Inserts fit in standard open racks and spindle racks in SteamScrubber and FlaskScrubber Glassware Washers. Holds 144 each 15 to 18 mm OD tubes. Grid size is 23.8 mm x 23.8 mm. For use in lower rack only.Specifications
Each | |
Type 304 Stainless Steel |