Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ SteamScrubber™ Glassware Washer

Clean glassware is essential in any lab. Labconco SteamScrubber™ Glassware Washer provides thorough, efficient, and economical cleaning that meets demanding laboratory standards.

Manufacturer:  Labconco™ 4400421

 View more versions of this product

Catalog No. 10-359-105


  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Add to cart

Specifications Description & Specifications

Specifications

27.5 in.
Ideal for college and university, government, pharmaceutical and industrial labs or any facility utilizing reusable glassware
230V
Domestic/international
4.7L/min. for hot tap water, 3.4L/min. for purified water
86.9 to 91.9cm
199°F
140°F
49°C
Type 304 Stainless Steel
0psi
13L/Fill
107kg
61.5cm
69.9cm
50/60Hz
12A
230V 50/60Hz
34.2 to 36.2 in.
Two open rack
120°F
93°C
60°C
20 to 120psi for hot tap water pressure at washer
SteamScrubber™ Glassware Washer
235 lb.
24.2 in.
UL 61010-1, CAN CSA C22.2 No. 61010.1, ETL, CE
Accessories
Promotions

Promotions

  • Labconco CorporationSupplier Diversity Partner
    Purchase any Labconco™ FlaskScrubber™ or SteamScrubber™ Glassware Washer and receive both a Utensil Basket and Utensil Holder at no additional cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.

    06-30-2018

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit