Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ SteamScrubber™ Glassware Washer

Clean glassware is essential in any lab. Labconco SteamScrubber™ Glassware Washer provides thorough, efficient, and economical cleaning that meets demanding laboratory standards.

Manufacturer:  Labconco™ 4400320

 View more versions of this product

Catalog No. 10-359-100


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

27.4 in.
Ideal for college and university, government, pharmaceutical and industrial labs or any facility utilizing reusable glassware
115V
Domestic
4.7L/min. for hot tap water, 3.4L/min. for purified water
86.6 to 91.7cm
180°F
140°F
49°C
Type 304 Stainless Steel
0psi
13L/Fill
93kg
61.2cm
69.6cm
60Hz
16A
115V 60Hz
34.1 to 36.1 in.
Two open rack
120°F
82°C
60°C
20 to 120psi for hot tap water pressure at washer
Undercounter SteamScrubber™
206 lb.
24.1 in.
UL 61010-1, CAN CSA C22.2 No. 61010.1, ETL
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit