Learn More
Labconco™ SteamScrubber™ Glassware Washer
Clean glassware is essential in any lab. Labconco SteamScrubber™ Glassware Washer provides thorough, efficient, and economical cleaning that meets demanding laboratory standards.
Manufacturer:  Labconco™ 4400331
 View more versions of this product
Catalog No. 10-359-103
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications Description & Specifications
Specifications
12A
230V 50/60Hz
34.1 to 36.1 in.
Two open rack, glass viewing window, 25w interior light
120°F
93°C
60°C
20 to 120psi for hot tap water pressure at washer
Undercounter SteamScrubber™
206 lb.
24.1 in.
UL 61010-1, CAN CSA C22.2 No. 61010.1, ETL, CE
69.6cm
50/60Hz
Domestic/international
4.7L/min. for hot tap water, 3.4L/min. for purified water
86.6 to 91.7cm
199°F
140°F
49°C
Type 304 Stainless Steel
0psi
13L/Fill
93kg
61.2cm
27.4 in.
Ideal for college and university, government, pharmaceutical and industrial labs or any facility utilizing reusable glassware
230V
Accessories