Learn More
Labconco™ SteamScrubber™ Labware Inserts
Hold a variety of wider-mouth flasks, beakers and other labware in inverted position
Manufacturer:  Labconco™ 4401501
 View more versions of this product
Catalog No. 15-352-808A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
8.8 in.
Each
FlaskScrubber™, SteamScrubber™ glassware washer
Type 304 Stainless Steel, Plastic
10.0
2.5 lb.
19.3 in.
22.35cm
Hold a variety of wider mouth flasks, beakers and other labware in inverted position
8 in.
20.3cm
Labware Insert
1.1kg
49cm