Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Support Shelves: Telescoping Base Stand

Manufacturer:  Labconco™ 3850100

 View more versions of this product

Catalog No. 10-269-259


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

10.4 in.
56cm
Support Shelf
Provides work surface to support remote equipment such as a circulator or the Aspirator Pump System within easy access of biosafety cabinet
Epoxy Coated Steel
10.4kg
26cm
22 in.
Purifier Logic Telescoping Base Stand
Epoxy-coated steel shelf and fasteners
23 lb.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit