Learn More
Labconco™ Telescoping Base Stands with Leveling Feet and Shelf (non-welded)
For use with Purifier™ Class II Biosafety Cabinets manufactured since 2000 including the Purifier Delta™, Logic™, Cell Logic™, Logic™+ and Cell Logic™+
Supplier: Labconco™ 3401004
Description
- Made of rugged powder coated tube and steel
- Provides work surface of 27.5 x 34.5 in. with 1 in. increments
- Includes leveling feet and shelf
Some assembly required
Specifications
27.5 to 34.5 in. | |
56.7 Kg | |
50.5 in. | |
128.3 cm |
125 lb. | |
29 in. | |
73.7 cm |