Learn More
Labconco™ Three-Way Valve for CentriVap™ System
For use with CentriVap™ Concentrators.
Manufacturer:  Labconco™ 7474000
Catalog No. 16-107-198
Add to cart
Description
Description
For use with CentriVap™ Concentrators.
Three-Way Valve allows connection of other laboratory equipment to CentriVap System, polypropylene, 0.5 in. OD.
Specifications
Specifications
CentriVap™ benchtop and mobile system concentrator
0.5 in.
Polypropylene
0.5 kg
1 lb.
1.27 cm
Three-way Valve