Learn More
Labconco™ Ultraviolet Lamp Kit
For Purifier™ Logic™+ Biosafety Cabinets and PuriCare™ Procedure Stations
Manufacturer: Labconco™ 3858502
Description
Ultraviolet Lamp Kits include 254 nm UV lamp and ballast. For use with Labconco Purifier Logic+ and Axiom Biosafety Cabinets and PuriCare Procedure Stations. Not recommended for Cell Logic+ Biosafety Cabinets with Scope-Ready package. Installation is required.Specifications
72 in. | |
254nm UV lamp, installation instructions and ballast | |
2.3kg |
182cm | |
5 lb. | |
All 230V models |