Learn More
Labconco™ UV Light for Purifier™ Delta Biosafety Cabinets
UV lamps for Labconco Purifier Biosafety Cabinets and PuriCare Procedure Stations.
$131.00 - $227.00
Specifications
For Use With (Equipment) Purifier Delta Biosafety Cabinet
View More Specs
Products 2
Catalog Number Mfr. No. Size Weight (English) Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size Weight (English) Weight (Metric) Price Quantity & Availability  
09-039-066
View Documents
Labconco™
1231900
6 ft. 17 lb. 7.7kg Each for $227.00
Add to cart
09-040-125
View Documents
Labconco™
1271300
3 ft. 5 lb. 2.3kg Each for $131.00
Add to cart
Description
Description
UV lamp for use with 3', 4', 5' and 6' Purifier Axiom, Logic+, Cell Logic+ and Delta Biosafety Cabinets and PuriCare Procedure Stations.
Specifications
Specifications
Purifier Delta Biosafety Cabinet