Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ XPert™ Balance Enclosures - Tall, 3 Ft. Wide

Enclosures protect user by keeping powders, particulates and fumes contained

Manufacturer:  Labconco™ 3930331

 View more versions of this product

Catalog No. 10-090-263


  Check Availability
Add to cart

Requires : FilterMate Portable Exhauster(s) or remote blower, work surface, supporting base, ductwork—all sold separately. Contact your customer service representative for more information.

Specifications Description & Specifications

Specifications

23.4 in.
29 x 3 x 32 in. (73.7 x 91.4 x 81.3cm)
New_Flow Rate
10 in.
Guardian™ Jr. airflow monitor
165 lb.
75kg
91.4cm
2
29 in.
ANSI Z9.5, ASHRAE 110, SEFA 1
32 in.
81.3cm
60Hz
73.7cm
115V
34.8 in.
88.4cm
23.4 x 34.8 x 31.5 in. (59.4 x 88.4 x 80cm)
115V 60Hz
Epoxy Coated Aluminum
24.4cm
Enclosure
165 lb.
75kg
37.8 in.
36 in.
73.6cm
59.4cm
31.5 in.
80cm
29 in.
FilterMate Portable Exhauster(s) or remote blower; work surface; supporting base; ductwork.
3 ft.
91.4cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit