Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ XPert™ Bulk Powder Enclosures

Exhausts hazardous airborne particulates and fumes to outside

Manufacturer:  Labconco™ 3963601

 View more versions of this product

Catalog No. 10-265-161


  Check Availability
Add to cart

Requires : Venting; can be connected to a remote blower or house exhaust and exhausted to the outside.

Specifications Description & Specifications

Specifications

115V 60Hz
12 in.
60Hz
115V
93.98cm
30 in.
76.2cm
Enclosure, with access opening center
191.7cm
1000 lb.
179.8 cm
30.5cm
72 in.
Fluorescent
37 in.
182.8cm
75.5 in.
Stainless Steel
32 in.
2 Utility
70.8 in.
SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit