Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco XPert Bulk Powder Filtered Systems
Protects from hazardous airborne particulates and fumes generated during powder transfer operations
$29413.00 - $33588.00
Specifications
Compliances | SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE |
---|---|
Depth (English) Interior | 30 in. |
Depth (Metric) Interior | 76.2cm |
Depth (English) | 37 in. |
Depth (Metric) | 93.98cm |
Catalog Number | Mfr. No. | Hertz | Voltage | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Hertz | Voltage | Price | Quantity & Availability | ||||
10-265-143
|
Labconco
3961521 |
50/60Hz | 230V |
Each for $30,062.00
|
|
||||
10-265-144
|
Labconco
3961600 |
60Hz | 115V |
Each for $32,938.00
|
|
10-265-141
|
Labconco
3961501 |
60Hz | 115V |
Each for $29,414.00
|
|
10-265-140
|
Labconco
3961500 |
60Hz | 115V |
Each for $29,414.00
|
|
10-265-142
|
Labconco
3961520 |
50/60Hz | 230V |
Each for $30,062.00
|
|
10-265-145
|
Labconco
3961601 |
60Hz | 115V |
Each for $32,938.00
|
|
10-265-146
|
Labconco
3961620 |
50/60Hz | 230V |
Each for $33,588.00
|
|
10-265-147
|
Labconco
3961621 |
50/60Hz | 230V |
Each for $33,588.00
|
|
||||
10-304-153
|
Labconco
3961502 |
60Hz | 115V |
Each for $29,413.00
|
|
10-304-155
|
Labconco
3961602 |
60Hz | 115V |
Each for $32,938.00
|
|
10-304-156
|
Labconco
3961622 |
50Hz | 230V |
Each for $33,588.00
|
|
10-304-154
|
Labconco
3961522 |
50/60Hz | 230V |
Each for $30,062.00
|
|
||||
Description
- Clean-Sweep openings and a zone-perforated rear baffle enhance horizontal laminar airflow
- With built-in Guardian 1000 digital airflow monitor
- Upper dilution air supply with by-pass air
- Upper containment sash foil and side-entry air foils
- Stainless steel work surface with front spill trough
- Transfer caddy with caddy latch and positioning plate
- Push-button operated electric hydraulic lift mechanism for raising the container to the optimal ergonomic height
- 30 in. interior depth and 32 in. interior height
- Tempered safety glass front sash and sides
- 12 in. high sash opening; ergonomic angled sash with sealed gas-assisted springs hold sash open (28 in. loading height)
- Built-in blower; outside ducting not required
- 99.99% efficient HEPA filter
- True bag-in/bag-out filter disposal system
- Pre-wired fluorescent light and switches for remote blower and light
- Epoxy-coated steel exterior
- 2 in. square-tube epoxy-coated steel base stand
- Left-side mounted waste chute
- Fourplex electrical receptacle and two utility ports
Compliance
SEFA 1-2002, Modified ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1
Specifications
SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE | |
76.2cm | |
93.98cm | |
Stainless Steel | |
210.8cm | |
2 Utility | |
30.5cm |
30 in. | |
37 in. | |
83 in. | |
32 in. | |
Fluorescent | |
12 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title