Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ XPert™ Bulk Powder Filtered Systems

Protects from hazardous airborne particulates and fumes generated during powder transfer operations

Manufacturer:  Labconco™ 3961501

 View more versions of this product

Catalog No. 10-265-141


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

115V 60Hz
12 in.
60Hz
115V
93.98cm
30 in.
76.2cm
Fume Hood
210.8cm
2 Utility
SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE
30.5cm
60 in.
Fluorescent
37 in.
152.4cm
83 in.
Stainless Steel
32 in.
Center Access Opening
965 lb.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit