Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Xpert™ TXE™ Equipment Enclosure

Labconco Xpert™ TXE™ Equipment Enclosure provides a safe and ventilated space to keep powder-generating apparatus.

Manufacturer:  Labconco™ 3892000

 View more versions of this product

Catalog No. 13-118-50


  Check Availability
Add to cart

Requires : Telescoping Base Stand

Specifications Description & Specifications

Specifications

HEPA
144cm
Fluorescent
ANSI Z9.5, CAN/CSA C22.2, ETL, ASHRAE 110, SEFA 1, UL
115V 60Hz
42 in.
60Hz
61cm
250 lb.
31 in.
78.7cm
56.9 in.
24 in.
Epoxy Coated Metal
24 x 31 x 56.9 in. (61 x 78.7 x 144cm)
Pharmaceutical research and quality control laboratories
106.6cm
w/o Guardian Airflow Monitor
115V
113kg
24 in.
61cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit