Learn More
General Data Healthcare™ LabCycle Specimen Tracking Software
Streamline lab workflow efficiency and eliminate errors
$37850.00 - $181000.00
Specifications
Description Applications: Labeling for specimen requisitions, containers, cassettes and slides
View More Specs
Products 4
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
22-048-211
View Documents
General Data Healthcare™
G17DDLCT1
Tracking Software T1
Each for $37,850.00
Add to cart
22-048-212
View Documents
General Data Healthcare™
G17DDLCT2
Tracking Software T2
Each for $74,550.00
Add to cart
22-048-213
View Documents
General Data Healthcare™
G17DDLCT3
Tracking Software T3
Each for $99,050.00
Add to cart
22-048-214
View Documents
General Data Healthcare™
G17DDLCENT
Tracking Software ENT
Each for $181,000.00
Add to cart
Description
Description
LabCycle is a scalable software solution used to identify specimens throughout the histopathology laboratory’s processes.
User-Friendly:
  • Customizable prompts allow specific laboratory terms to be utilized
  • Intuitive user screens make it easier to learn the software minimizing the training period
  • Role-based access allows users to only view their assigned modules
Scalable:
  • Modules can be implemented in phases
  • Laboratories decide how much of system to implement
  • Supports multiple sites
Specimen Information:
  • Real-time access to a specimen’s current location
  • Ability to view a specimen’s history
  • Tracking of instruments used to process the specimen
Dashboard and Reports: 
  • Easily view the number of specimens processed
  • View key performance indicators (KPIs) such as turnaround time (TAT)
  • Locate specimens that have not moved throughout the laboratory within an expected timeframe
Hardware Compatibility:
  • Option to utilize existing modern computer systems (meeting minimum system requirements)
  • Native support for existing ID/Positive barcode scanners, slide labelers and laser cassette markers
  • Ability to use compatible barcode scanners, slide labelers and cassette printers
Application: Labeling for specimen requisitions, containers, cassettes and slides
Specifications
Specifications
Applications: Labeling for specimen requisitions, containers, cassettes and slides