Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Brady™ RTK Pictogram Labels, Corrosive
Effectively convey personal protection instructions and special precautionary information. Brady™ RTK Pictogram Labels are made from premium grade B-946 vinyl and are sized to fit standard blank write-on right-to-know labels.
Manufacturer:  Brady™ 58585
 View more versions of this product
Catalog No. 19-107-235
Description
Description
  • Made from premium grade B-946 vinyl
  • Sized to fit standard Blank Write-On Right-To-Know Labels
  • Correspond to ANSI proposed Z.535 Standard
  • Card-mounted for easy removal
Specifications
Specifications
Acrylic, Cold Temperature, Permanent
W x H: 0.75 x 0.75 in.
Pressure Sensitive Vinyl (B-946)
Blue, White
0.004 in.
RTK Pictogram Labels, Corrosive