Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazardous Material Shipping Labels: DANGER: DO NOT LOAD IN PASSENGER AIRCRAFT CARGO AIRCRAFT ONLY…

Use our Hazardous Material Shipping Labels to insure all domestic and foreign hazardous material shipments reach their destinations without costly delays or rejections

Manufacturer:  Brady™ 63239

Catalog No. 19-107-327


Add to cart

Description

Description

  • Vinyl, Black on Orange, 4.313 in H x 4.75 in W
Specifications

Specifications

Self-Adhesive
Permanent Acrylic
Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704; OSHA 1910.1200
Label
Gloss
Each
1
4.313H x 4.75W x 0.005 in. D
148.59 mm
Vinyl
Each
Not Autoclavable
Black on Orange
0.005 in.
4.313 in. H x 4.75 in. W x 0.005 in. D
4.313 in.
Each
Rectangle
4.75 in.
English
Label
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit