Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Hazardous Material Shipping Labels: Blue on White, MAGNETIZED MATERIAL KEEP AWAY FROM AIRCRAFT COMPASS DETECTOR UNIT
Blue on White, 3.563 H x 4.313 in W
Supplier: Brady™ 63241
Description
Made of plasticSpecifications
Not Autoclavable | |
Label | |
Blue on White | |
Plastic | |
Label | |
1 Ea. | |
4.313 x 3.563 in. (W x H) | |
4.313 in. | |
3.563 in. |
Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704; OSHA 1910.1200 | |
Self-adhesive | |
Label | |
One | |
English | |
Rectangle | |
3.563 x 4.313 in. | |
133.35 mm |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction