Learn More
Mirus Bio™ Label IT™ Nucleic Acid Labeling Kit, Digoxin
Label IT™ Nucleic Acid Labeling Reagents for efficient, direct, non-enzymatic labeling of DNA and RNA.
Supplier:  Mirus Bio™ MIR 3325
 View more versions of this product
Catalog No. MIR3325
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Label Any DNA or RNA Template – Suitable for a wide range of applications
  • One-step Chemical Method – Easily and consistently control the labeling reactions
  • Adjustable Labeling Density – Achieve high sensitivity with optimally labeled DNA and RNA
  • Covalent Mechanism – Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications; labels do not impact hybridization performance
NOTE: All the Label IT Reagents use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.
Specifications
Specifications
Chemiluminescent
Not Applicable
DNA/RNA
Label Reagent
Lyophilized
Lyophilized
DNA (Labeled), RNA (Labeled)
DNA/RNA
25 μg
Recommendations
Recommendations