Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady Hazardous Waste Label: NON-HAZARDOUS WASTE
Identify hazardous waste materials for transportation off-site. Brady Hazardous Waste Labels are made of vinyl and comply with all current federal regulations regarding identification of hazardous waste material transportation.
Manufacturer: Brady 121154
Description
- To identify containers, cylinders and drums holding hazardous waste
- Made of B-7569 vinyl
- Available in size (W x H): 15.2 x 15.2cm (6 x 6 in.)
Specifications
Hazardous Waste Label | |
Green, White | |
Square | |
0.004 in. | |
100/Pk. |
Acrylic, Permanent | |
NON-REGULATED WASTE | |
6 x 6 in. | |
Vinyl (B-7569) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title