Learn More
Labplas Filtra-Bag Blender Bags
Designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid/solid substances. Labplas Filtra-Bag Blender Bags are constructed with a polyethylene outer shell that reduces the risk of piercing the bag wall during blending.
Manufacturer:  Labplas EFT1015A
 View more versions of this product
Catalog No. 01-815-392
Add to cart
Description
Description
Designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid/solid substances. Labplas Filtra-Bag Blender Bags are constructed with a polyethylene outer shell that reduces the risk of piercing the bag wall during blending.
  • Used for aliquoting
  • Blender accessible
  • Filter membrane for separating solids and liquids
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
Specifications
Specifications
Safety tabs
Aliquoting
Sterile
10 in.
2720mL
PE (Polyethylene)
15 x 10 in. (381 x 381mm)
15 in.
3 mil
92 oz.
381mm
381mm