Learn More
Labplas Filtra-Bag Blender Bags
Designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid/solid substances. Labplas Filtra-Bag Blender Bags are constructed with a polyethylene outer shell that reduces the risk of piercing the bag wall during blending.
Manufacturer: Labplas EFT1515A
Description
Designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid/solid substances. Labplas Filtra-Bag Blender Bags are constructed with a polyethylene outer shell that reduces the risk of piercing the bag wall during blending.
- Used for aliquoting
- Blender accessible
- Filter membrane for separating solids and liquids
- Contaminant-free
- Virgin polyethylene
- Large bag mouth opening
- Easy to use pull tabs
Specifications
Safety tabs | |
Aliquoting | |
Sterile | |
15 in. | |
4000mL | |
PE (Polyethylene) |
15 x 15 in. (381 x 381mm) | |
15 in. | |
3 mil | |
138 oz. | |
381mm | |
381mm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.