Learn More
Labplas Filtra-Bag Blender Bags Available on GSA/VA Contract for Federal Government customers only.
Designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid/solid substances. Labplas Filtra-Bag Blender Bags are constructed with a polyethylene outer shell that reduces the risk of piercing the bag wall during blending.
Manufacturer:  Labplas SCT7012A
 View more versions of this product
Catalog No. 01-815-387
Add to cart
Description
Description
Designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid/solid substances. Labplas Filtra-Bag Blender Bags are constructed with a polyethylene outer shell that reduces the risk of piercing the bag wall during blending.
  • Used for aliquoting
  • Blender accessible
  • Filter membrane for separating solids and liquids
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Sterile
7 in.
1650mL
PE (Polyethylene)
Tear-off protection strip
Aliquoting
3 mil
55 oz.
305mm
305mm
12 x 7 in. (305 x 305mm)
12 in.