Learn More
Labplas Sani-Sponge Kit
The simpler solution to surface sampling
Manufacturer:  Labplas KSS61100
Catalog No. 01-002-71
Add to cart
Description
Description
Designed to collect samples to detect the presence of microbiological contaminations such as Listeria, Salmonella, E. coli, and other food-borne pathogens on almost any surface. Kit is convenient and easy to use, and helps eliminate several time-consuming steps.
  • All Labplas SANI-SPONGE TWIRL'EM™ bags come with safety tabs
  • Puncture-proof tabs eliminate the sharp points that can cause bag puncture and/or abrasions
  • Kits are sterilized using gamma irradiation
  • Produced in accordance with HACCP, USDA, ACIA and CFIA requirements
  • Available in Dry Sponge or Wet Sponge forms
Dry Sponge
  • TWIRL'EM™ Sterile Sampling bag with write-on strip containing a dry biocide-free cellulose sponge (1.5 in. x 3 in. x 0.625 in. when hydrated)
Wet Sponge
  • TWIRL'EM Sterile Sampling bag accompanied by a foil-sealed, pre-moistened cellulose sponge, holding 10mL of neutralizing buffer-sterile
Specifications
Specifications
9 x 4.5 in. (229 x 114mm)
3 mil
18 oz.
229mm
114mm
9 in.
4.5 in.
540mL
Sponge