Learn More
Labplas Sani-Sponge Sampling Bags, Regular tabs: With Clear Body, Tear-Off Protection Strip Available on GSA/VA Contract for Federal Government customers only.
Designed to collect samples to detect the presence of microbiological contaminations
Manufacturer:  Labplas KSS67110DE
 View more versions of this product
Catalog No. 01-815-380
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Detects Listeria, Salmonella and E. coli
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
  • Sponge for swabbing
  • Dehydrated sponge
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
DE Neutralizing buffer
15 oz.
PE (Polyethylene)
2.5 mil.
450mL