Learn More
Labplas Sani-Sponge Sampling Bags, Regular tabs: With Gloves, Tear-Off Protection Strip
Designed to collect samples to detect the presence of microbiological contaminations
Manufacturer:  Labplas KSS61105
 View more versions of this product
Catalog No. 01-815-377
Add to cart
Description
Description
  • Detects Listeria, Salmonella and E. coli
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
  • Sponge for swabbing
  • Dehydrated sponge
Specifications
Specifications
Dehydrated sponge with gloves
2.5 mil.
PE (Polyethylene)