Learn More
Labplas Secure-T Sample Bags Available on GSA/VA Contract for Federal Government customers only.
Made to handle the most robust blending applications
Manufacturer:  Labplas SCL41214
 View more versions of this product
Catalog No. 01-815-396
  Check Availability
Add to cart
Description
Description
  • Durable
  • Can with stand blender use
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
  • Open top
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Tear-off protection strip
4 mil
PE (Polyethylene)
356 x 305mm (14 x 12 in.)
4500mL