Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labplas Twirl'EM™ Environmentally Friendly Sterile Sampling Bags, SafetyTabs: Clear Body Closure W 1 Round Wire and 1 Flat Wire

An economical and timesaving green alternative to cleaning and storing reusable rigid containers

Manufacturer:  Labplas EFL47012E

 View more versions of this product

Catalog No. 01-815-352


Add to cart

Description

Description

  • Environmentally friendly
  • Sterile
  • Fully biodegradable LDPE plastic tubing
  • Can be heated to 240°C for sterility
  • Large bag mouth opening
  • Easy pull tabs to open and close the bag

Specifications

Specifications

Closure with one round and one flat wire
12 x 7 in. (305 x 178mm)
4 mil
55 oz.
305mm
178mm
Clear body
12 in.
7 in.
1650mL
PE (Polyethylene)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit