Learn More
Labplas Twirl'EM™ Environmentally Friendly Sterile Sampling Bags, SafetyTabs: Printed Body Closure With 2 Round Wires
An economical and timesaving green alternative to cleaning and storing reusable rigid containers
Specifications
Closure Type Closure with one round and one flat wire
Material PE (Polyethylene)
Description Printed body
View More Specs
Description
Description
  • Environmentally friendly
  • Sterile
  • Fully biodegradable LDPE plastic tubing
  • Can be heated to 240°C for sterility
  • Large bag mouth opening
  • Easy pull tabs to open and close the bag
Specifications
Specifications
Closure with one round and one flat wire
PE (Polyethylene)
Printed body