Learn More
Labplas Twirl'EM™ Large Format Sampling Bags with Tabs Available on GSA/VA Contract for Federal Government customers only.
Polyethylene sampling bags with large openings
Manufacturer:  Labplas EDL41220
 View more versions of this product
Catalog No. 01-815-367
Add to cart
Description
Description
  • Large sized
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Regular tabs
Sterile
8000mL
Regular Tabs
4 mil
PE (Polyethylene)