Learn More
Labplas Twirl'EM™ Large Format Sampling Bags with Tear-Off Protection Strip Available on GSA/VA Contract for Federal Government customers only.
Large format bags with tear-off protection strip
Manufacturer:  Labplas SCL43048
 View more versions of this product
Catalog No. 01-815-373
Add to cart
Description
Description
  • Large sized
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
125/Cs.
2600 oz.
Color
Large Format Sampling Bags with Tear-Off Protection Strip
2 mil
81000mL
Virgin polyethylene
122 x 76cm (48 x 30 in.)