Learn More
Labplas Twirl'EM™ Large Format Sampling Bags with Tear-Off Protection Strip
Large format bags with tear-off protection strip
Manufacturer:  Labplas SCL21520
 View more versions of this product
Catalog No. 01-815-371
Add to cart
Description
Description
  • Large sized
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
Specifications
Specifications
500/Cs.
405 oz.
Color
Large Format Sampling Bags with Tear-Off Protection Strip
4 mil
12000mL
Virgin polyethylene
50.8 x 38.1cm (20 x 15 in.)