Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Labplas Twirl'EM™ Stand Up Sampling Bags, Safety tabs Available on GSA/VA Contract for Federal Government customers only.
Meant for jobs where two hands are required
Manufacturer:  Labplas EPS5590
 View more versions of this product
Catalog No. 01-815-375
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Stand upright by themselves
  • No rack or stand needed
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
600mL
PE (Polyethylene)