Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labplas Twirl'EM Sterile Sampling Bags, SafetyTabs
Provide a sterile, secure, contaminant-free pliant container that ensures dependable analysis results. Labplas Twirl'EM Sterile Sampling Bags with safety tabs incorporate easy-to-pull tabs that open the bag without risk of contaminating the bag interior. They are useful in a variety of sampling applications, including environmental and surface sampling.
Manufacturer: Labplas EPR5590
Description
- Virgin polyethylene tubing
- Can be heated to 240°C for sterility
- Large bag mouth opening to facilite insertion of samples
- Easy pull tabs to open and close the bag with decreased risk of contamination
- R-Nase, D-Nase, Pyrogen, and BPA free
- Enviromental sampling
- Surface sampling
- Carcass sampling
- Biomedical research
- Pharmaceutical research
- Food industry
Specifications
Sterile | |
650mL | |
Closure with two round wires |
3 mil | |
PE (Polyethylene) | |
Printed body |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title