Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labplas Twirl'EM™ Sterile Sampling Bags, SafetyTabs

Provide a sterile, secure, contaminant-free pliant container that ensures dependable analysis results. Labplas Twirl'EM™ Sterile Sampling Bags with safety tabs incorporate easy-to-pull tabs that open the bag without risk of contaminating the bag interior. They are useful in a variety of sampling applications, including environmental and surface sampling.

Manufacturer:  Labplas EFL1015

 View more versions of this product

Catalog No. 01-815-325


  Check Availability
Add to cart

Description

Description

  • Virgin polyethylene tubing
  • Can be heated to 240°C for sterility
  • Large bag mouth opening to facilite insertion of samples
  • Easy pull tabs to open and close the bag with decreased risk of contamination
  • R-Nase, D-Nase, Pyrogen, and BPA free
Applications include:
  • Enviromental sampling
  • Surface sampling
  • Carcass sampling
  • Biomedical research
  • Pharmaceutical research
  • Food industry
Specifications

Specifications

Closure with one round and one flat wire
Sterile
4000mL
Clear body
4 mil
PE (Polyethylene)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit