Learn More
Labplas Water Sampling Kits, Regular tabs Available on GSA/VA Contract for Federal Government customers only.
Designed to collect, carry and contain chlorinated water samples in aseptic conditions and are an economical and efficient alternative to rigid sampling containers
Manufacturer:  Labplas KWS22200
 View more versions of this product
Catalog No. 01-815-384
  Check Availability
Add to cart
Description
Description
  • Designed to collect and carry chlorinated water samples
  • Sterile kit contains non-toxic meatralizing tablets of sodium thiosulfate
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
300mL
PE (Polyethylene)