Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ LabServ Microcentrifuge Tubes with Standard Snap Caps
Microtubes feature standard snap caps in choice of clear or multicolored vials
Manufacturer:  Thermo Scientific™ 502GRDMSERVCONV
 View more versions of this product
Catalog No. 02-707-513
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Withstand forces up to 30,000 x g
  • Temperature range of -80° to +121°C
  • Available in either natural or mixed colors
  • RNase, DNase-free
Specifications
Specifications
Snap cap
0.5mL
Mixed
30000xG