Learn More
Fisherbrand™ LabServ Microcentrifuge Tubes with Standard Snap Caps
Microtubes feature standard snap caps in choice of clear or multicolored vials
Manufacturer:  Fisherbrand™ 508GRDMSERVCONV
 View more versions of this product
Catalog No. 02-707-517
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Withstand forces up to 30,000 x g
  • Temperature range of -80° to +121°C
  • Available in either natural or mixed colors
  • RNase, DNase-free
Specifications
Specifications
Snap cap
2mL
Mixed
30000xG