Learn More
Thermo Scientific™ Labserv Polypropylene Centrifuge Tubes
Sterile tubes with graduated divisions
Manufacturer:  Thermo Scientific™ 14955161
 View more versions of this product
Catalog No. 14-955-161
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
  • Made from polypropylene
  • DNase/RNase and pyrogen free
  • RCF rating of 12,000xG
  • Available in rack or sleeve
Description & Specifications
Specifications
2.5mL
DNase/RNase and pyrogen free
Yes
Centrifuge Type
12,000xG
Polypropylene
Rack