Learn More
Thermo Scientific™ Labtainer™ BioProcess Container 2D System, ASI 28 Film, 3 Edge Ports, MPC Insert/Body with C-Flex Tubing/Pinch Clamps
Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.
$1370.00 - $1665.00
Specifications
Port Detail MPC insert and body connectors and injection site
Tubing Style C-Flex, EVA
Sterilization Method Irradiated
Material Ethylene Vinyl Acetate (EVA)
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
SS00020I View Documents Thermo Scientific™
SS00020I
5 L Case of 30 for $1,665.00
Add to cart
SS00021I View Documents Thermo Scientific™
SS00021I
10 L Case of 25 for $1,443.75
Add to cart
SS00022I View Documents Thermo Scientific™
SS00022I
20 L Case of 25 for $1,518.75
Add to cart
SS00023I View Documents Thermo Scientific™
SS00023I
50 L Case of 20 for $1,370.00
Add to cart
Description
Description
This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.
  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors
Specifications
Specifications
MPC insert and body connectors and injection site
C-Flex, EVA
Irradiated
Ethylene Vinyl Acetate (EVA)