Learn More
InVitriaSupplier Diversity Partner LACROMIN HOLO RLACTOFERRIN 10G DFS Item
Manufacturer:  InVitriaSupplier Diversity Partner 777LAC01510G
Lacromin, recombinant human lactoferrin is an animal component free cell culture media supplement that increases cell growth while protecting cells from apoptosis (programmed cell death). Research has demonstrated that Lacromin often outperforms and is used as a replacement for more expensive Insulin-like Growth Factor (IGF). Lacromin is a simple, yet powerful tool used to optimize media for maximum cell viability and growth. As a recombinant lactoferrin, Lacromin provides animal-free, defined, and consistent performance. Advantages: animal component free, improves cell growth, replaces more expensive IGF growth factor, protects cells from apoptosis, well defined and consistent, regulatory friendly.
Catalog No. NC9953247