LaCrosse Insulated Storm Boots

Manufacturer: LACROSSE SAFETY & INDUSTRIAL 10905S09

 View more versions of this product

Catalog No. 19-817-462

  Check Availability
Add to Cart

    Specifications

    Liner Material Canvas
    Certifications/Compliance ASTM F2413-11 M I/75 C/75 EH PR
    For Use With (Application) Cold, muddy wet jobsites
    Height (English) Heel 0.31 in.
    Height (English) 32 in.
    Height (Metric) Heel 0.8cm
    Resistant Ozone and abrasion resistant
    Sole Features Midsole, Trac-lite outsole, Contoured heel, 8mm Wool felt insole
    Toe Material Steel
    Color Black
    Material Rubber
    Size 9

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit