LaCrosse Quad Comfort 4 x 8 Steel Toe Work Boots

Manufacturer: LACROSSE SAFETY & INDUSTRIAL 480002 4.5M

 View more versions of this product

Catalog No. 19-077-757

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Sole Features Rubber, slip resistant Quad outsole
    Size 4.5M
    Certifications/Compliance ASTM F2413-05 M I/75 C/75 EH
    For Use With (Application) General purpose work
    Gender Unisex
    Liner Material Hyper-dri™
    Shank Material Nylon
    Toe Material Steel
    Tread Style Smooth minimal
    Waterproof Yes
    Weight (English) 4 lb.
    Color Brown
    Eyelets Material Metal
    Height (English) 8 in.
    Weight (Metric) 1.81kg
    Material Full Grain Leather

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit