Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

LaCrosse Quad Comfort 4 x 8 Steel Toe Work Boots

Manufacturer:  LACROSSE SAFETY & INDUSTRIAL 480002 16M

 View more versions of this product

Catalog No. 19-077-776

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Specifications

ASTM F2413-05 M I/75 C/75 EH
General purpose work
8 in.
Nylon
Steel
Yes
1.81kg
Full Grain Leather
Metal
Unisex
Hyper-dri™
Direct attach (chemically bonded)
Smooth minimal
4 lb.
Brown
16M
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit