Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactase Mouse anti-Human, Janelia Fluor 549, Clone: 3C105.1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250509JF549
Description
Lactase Monoclonal specifically detects Lactase in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Lactase | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LCT | |
Normal human jejunal epithelial brush border-enriched membranes from a non-secretor and blood group O | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
3C105.1 | |
Western Blot, Immunohistochemistry | |
EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1 | |
Mouse | |
Protein G purified | |
Cancer, Cellular Signaling, metabolism | |
3938 | |
Store at 4°C in the dark. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title