Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactate Dehydrogenase A/LDHA Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI2D11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271121AF647

Catalog No. N271121A647


Add to cart

Description

Description

Lactate Dehydrogenase A/LDHA Monoclonal antibody specifically detects Lactate Dehydrogenase A/LDHA in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Lactate Dehydrogenase A/LDHA
OTI2D11
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
LDHA
Full length recombinant protein of human Lactate Dehydrogenase A/LDHA (NP_005557) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Mitochondrial Mediated Pathway, Neuroscience
Primary
3939.0
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit