Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactate Dehydrogenase A/LDHA Rabbit anti-Human, Rat, Clone: 2066C, R&D Systems™

Rabbit Monoclonal Antibody

$125.35 - $329.00

Specifications

Antigen Lactate Dehydrogenase A/LDHA
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Format Purified
Clone 2066C
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB9158SP View Documents R&D Systems
MAB9158SP
25 ug Each for $125.35
Add to cart
 
MAB9158100 View Documents R&D Systems
MAB9158100
100 ug Each for $329.00
Add to cart
 
Description

Description

LDHA (lactate dehydrogenase A chain; also LDH-M and PIG-19) is a 34-36kDa member of the LDH family of enzymes. It is a part of a cytoplasmic complex that is found principally in hepatocytes and skeletal muscle. The LDHA complex catalyzes the conversion of pyruvate to lactate, thereby generating NAD, the final step in anerobic glycolysis. This NAD is essential for the subsequent generation of ATP. Human LDHA is 332 amino acids (aa) in length. It contains an N-terminal coenzyme binding region, a central catalytic site, and at least nine utilized Lys acetylation and two Tyr phosphorylation sites. It also undergoes ISGylation where a 17kDa product of the ISG15 gene is covalently attached to LDHA in a ubiquitin-like manner. LDHA forms tetramers composed of two dimers of varying composition. The tetramer may be homotetrameric (four monomers), or contain from one to three substitute LDHB monomers typically found in heart muscle. There are multiple splice variants. One possesses a five aa substitution for aa 237-332, a second contains a 45 aa substitution for aa 230-332, a third shows a deletion of aa 82-139, while a fourth utilizes an alternative start site 29 aa upstream of the standard site. Over aa 2-92, human LDHA shares 93% aa sequence identity with mouse LDHA.
Specifications

Specifications

Lactate Dehydrogenase A/LDHA
Purified
Immunocytochemistry, Immunohistochemistry, Western Blot
Rabbit
Protein A or G purified from hybridoma culture supernatant
RUO
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
2066C
Unconjugated
IgG
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit