Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lactate Dehydrogenase A/LDHA Rat Recombinant Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
$191.70 - $494.50
Specifications
Concentration | 0.25mg/mL |
---|---|
For Use With (Application) | SDS-PAGE |
Formulation | In PBS (pH 7.4) containing 1mM DTT, 20% glycerol |
Gene ID (Entrez) | 3939 |
Molecular Weight (g/mol) | TMW: 37.5kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP250564B
|
Novus Biologicals™
NBP25056410UG |
10μg |
Each for $191.70
|
|
|||||
NBP250564C
|
Novus Biologicals™
NBP25056450UG |
50μg |
Each for $494.50
|
|
|||||
Description
A bioactive recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-332 of Rat Lactate Dehydrogenase A/LDHA The Recombinant Rat Lactate Dehydrogenase A/LDHA Protein is derived from E. coli. The Recombinant Rat Lactate Dehydrogenase A/LDHA Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.Specifications
0.25mg/mL | |
In PBS (pH 7.4) containing 1mM DTT, 20% glycerol | |
TMW: 37.5kDa | |
Store at 4°C short term. Aliquot and store at −70°C long term. Avoid freeze-thaw cycles. | |
Lactate Dehydrogenase A/LDHA | |
Recombinant |
SDS-PAGE | |
3939 | |
Baculovirus | |
Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59 | |
Unconjugated | |
>95% pure by SDS-PAGE |
For research use only and is not approved for use in humans or in clinical diagnosis
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title