Learn More
LACTB2 Mouse anti-Canine, Human, Non-human primate, Clone: OTI2F9, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525465
Catalog No. PIMA525465
Add to cart
Description
Description
Endoribonuclease; cleaves preferentially 3 ft. to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3 ft. -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.
Specifications
Specifications
LACTB2
Monoclonal
1.1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q53H82
beta-lactamase-like protein 2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
486990, 51110, 695035
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2F9
Unconjugated
LACTB2
Liquid
LACTB2
Human recombit protein fragment corresponding to amino acids 78-288 of LACTB2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate